Learn More
IGF1R alpha Monoclonal Antibody (1H7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA181739
Additional Details : Weight : 0.01000kg
Description
Clone 1H7 recognizes an epitope in the alpha subunits of CD221, demonstrated by Western blotting, and is reported to partically block binding of IGF-I and IGF-II to CD221. This product requires protein digestion pre-treatment of paraffin sections using trypsin or pronase. A suggested positive control for immunohistochemical applications is pancreas or placenta. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. This product is a low-endotoxin, preservative-free formulation. Endotoxin level is <0.01 EU/ug.
IGF1R alpha Monoclonal antibody specifically detects IGF1R alpha in Human samples. It is validated for Flow Cytometry, Functional Assay, Immunohistochemistry (Paraffin), Western Blot.Specifications
IGF1R alpha | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
P08069 | |
Igf1r | |
Proprietary Immunogen. | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Functional Assay, Immunohistochemistry (Paraffin), Western Blot | |
1H7 | |
Unconjugated | |
Igf1r | |
A330103N21Rik; CD221; D930020L01; hyft; IGF-1 receptor; Igf1r; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; insulin-like growth factor 1 receptor beta subunit; insulin-like growth factor I receptor; insulin-like growth factor receptor 1; insulin-like growth factor-I receptor; insulin-like growth factor-I receptor precursor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2 | |
Mouse | |
Protein G | |
RUO | |
3480 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |