Få mere at vide
CD220 Mouse, Unlabeled, Clone: 46, BD
Beskrivelse
Insulin Receptor (IR) is a transmembrane receptor tyrosine kinase which, upon insulin binding, initiates a cascade of events, including autophosphorylation, phosphorylation of cellular protein substrates, glucose transport, and glycogen synthesis. IR is synthesized as a large glycosylated precursor that is cleaved upon maturation into a 130kDa α-subunit with kinase activity and a 95kDa β-subunit. The active Insulin Receptor is a heterotetramer of homologus α and β subunits joined by disulfide bonds. Among the major cytosolic substrates of the Insulin Receptor are IRS-1 and -2, β-Adrenergic receptor, and pp15 (adipocyte lipid-binding protein, ALBP). Autophosphorylation of the IR recruits IRS-1 and -2 to the phosphotyrosines. Subsequently, the phosphorylated IRS-1 and -2 act as docking sites for other signaling proteins like PI3-Kinase, Shc, PTP1D, Nck, etc. In addition, the phosphatase LAR is tightly associated with the IR and LAR becomes activated after insulin stimulation dephosphorylating the IR and its substrates. Therefore, LAR provides a turn-off mechanism in insulin signaling.
Host Species: Mouse
Clone: 46
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Human Insulin pro-receptor β subunit 1006-1144
Formula Weight [Chemical]: 95kDa
Immunofluorescence, Immunoprecipitation, Western Blotting
Tekniske data
Tekniske data
| Antigen | CD220 |
| Applikationer | Western Blot |
| Klassifikation | Monoclonal |
| Klon | 46 |
| Koncentration | 250μg/mL |
| Konjugeret | Unconjugated |
| Oversigt | Insulin Receptor β |
| Formulering | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Værtsarter | Mouse |
| Immunogen | Human Insulin pro-receptor β subunit 1006-1144 |
| Vis mere |
For Research Use Only.
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.