Learn More
CD206 Monoclonal Antibody (MR5D3), PE, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA516872
Additional Details : Weight : 0.01000kg
Description
CD206 is expressed weakly at the cell surface. Staining may be increased following membrane permeabilization. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. Rat anti mouse CD206 antibody, clone MR5D3 recognizes the mouse mannose receptor, a approximately 175 kDa type 1 membrane glycoprotein that is also known as CD206.
CD206 Monoclonal antibody specifically detects CD206 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD206 | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
Q61830 | |
Mrc1 | |
Chimaeric CRD4-7-Fc protein. | |
100 Tests | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry | |
MR5D3 | |
PE | |
Mrc1 | |
AW259686; bA541I19.1; CD206; CELE_Y39A3CR.4; CLEC13D; CLEC13DL; C-type lectin domain family 13 member D; C-type lectin domain family 13 member D-like; ddp-1; hMR; Human mannose receptor; macrophage mannose receptor 1; Macrophage mannose receptor 1-like protein 1; mannose receptor, C type 1; mannose receptor, C type 1-like 1; Mitochondrial import inner membrane translocase subunit Tim8; MMR; MR; MRC1; MRC1L1; tim-8; Y39A3CR.4 | |
Rat | |
Protein G | |
RUO | |
17533 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |