Learn More
CD200R Monoclonal Antibody (OX-102), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516895
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD200R Monoclonal antibody specifically detects CD200R in Rat samples. It is validated for Flow Cytometry.Specifications
CD200R | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
Q9ES58 | |
CD200R1 | |
Membrane fraction of thioglycollate-elicited rat peripheral cells. | |
100 Tests | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry | |
OX-102 | |
PE | |
CD200R1 | |
antigen identified by monoclonal antibody MRC OX-2 receptor; CD200 cell surface glycoprotein receptor; CD200 R1; CD200 receptor 1; CD200R; Cd200r1; Cell surface glycoprotein CD200 receptor 1; cell surface glycoprotein OX2 receptor 1; cell surface glycoprotein receptor CD200; CRTR2; HCRTR2; MOX2 receptor; MOX2R; Orexin receptor 2; OX102 antigen; OX2 receptor; OX2R; UNQ2522/PRO6015 | |
Mouse | |
Protein A | |
RUO | |
64357 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |