missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD200R Monoclonal Antibody (OX-102), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516894
2310.11 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD200R Monoclonal antibody specifically detects CD200R in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Specifications
CD200R | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Q9ES58 | |
CD200R1 | |
Membrane fraction of thioglycollate-elicited rat peripheral cells. | |
250 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
OX-102 | |
Unconjugated | |
CD200R1 | |
antigen identified by monoclonal antibody MRC OX-2 receptor; CD200 cell surface glycoprotein receptor; CD200 R1; CD200 receptor 1; CD200R; Cd200r1; Cell surface glycoprotein CD200 receptor 1; cell surface glycoprotein OX2 receptor 1; cell surface glycoprotein receptor CD200; CRTR2; HCRTR2; MOX2 receptor; MOX2R; Orexin receptor 2; OX102 antigen; OX2 receptor; OX2R; UNQ2522/PRO6015 | |
Mouse | |
Protein A | |
RUO | |
64357 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |