missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2 Monoclonal Antibody (OX-34), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA170018
1596.74 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
CD2 Monoclonal antibody specifically detects CD2 in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation.Specifications
CD2 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P08921 | |
CD2 | |
T blasts prepared in mixed lymphocyte reactions with purified rat T helper cells against irradiated spleen. | |
250 μg | |
Primary | |
Rat | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation | |
OX-34 | |
Unconjugated | |
CD2 | |
Cd2; CD2 antigen; CD2 antigen (p50), sheep red blood cell receptor; CD2 molecule; CD2R; cluster of differentiation 2; Erythrocyte receptor; FLJ46032; LFA2; LFA-2; LFA-3 receptor; Ly37; Ly-37; lymphocyte antigen 37; OX34; OX-34; OX-34 antigen; OX-45 surface antigen homolog to human T lymphocyte CD2 antigen; Rosette receptor; SRBC; surface immunoglobulin superfamily molecule; T11; T-cell surface antigen CD2; T-cell surface antigen T11/Leu-5 | |
Mouse | |
Protein G | |
RUO | |
497761 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |