missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2/CD7 Antibody Cocktail, FITC, PE, Invitrogen™
Antibody Cocktail
Brand: Invitrogen MA112004
2560.12 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
This antibody is a monoclonal mixture of FITC-conjugated CD2 (clone T6.3, isotype IgG2a) and PE-conjugated CD7 (clone MG34, isotype IgG2a).
CD2/CD7 Cocktail antibody specifically detects CD2/CD7 in Human samples. It is validated for Flow Cytometry.Specifications
CD2/CD7 | |
Cocktail | |
FITC/PE | |
CD7 | |
2CD2 antigen; CD2; CD2 molecule; CD7; CD7 antigen (p41); CD7 molecule; Erythrocyte receptor; GP40; LEU-9; LFA-2; LFA-3 receptor; p41 protein; Rosette receptor; SRBC; T11; T-cell antigen CD7; T-cell leukemia antigen; T-cell surface antigen CD2; T-cell surface antigen Leu-9; T-cell surface antigen T11/Leu-5; Tp40; TP41 | |
Mouse | |
Protein A/G | |
RUO | |
914, 924 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
T6.3, MG34, MG34 | |
PBS with 0.2% carrier protein and 0.08% sodium azide | |
P06729, P09564 | |
CD2, CD7 | |
T lymphocytes activated by mixed lymphocyte culture and T-acute lymphoblastic leukemia (T-ALL) cells | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a |