missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1a Monoclonal Antibody (HI149), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119605
Additional Details : Weight : 0.01000kg
Description
CD1a Monoclonal antibody specifically detects CD1a in Human samples. It is validated for Flow Cytometry.Specifications
CD1a | |
Monoclonal | |
FITC | |
CD1A | |
CD1; CD1 antigen; CD1.1; CD1A; CD1A antigen, a polypeptide; CD1a molecule; cd1a1; CD1A1 antigen; cluster of differentiation 1 A; cortical thymocyte antigen CD1A; differentiation antigen CD1-alpha-3; epidermal dendritic cell marker CD1a; FCB6; HTA1; hTa1 thymocyte antigen; R4; T6; T-cell surface antigen T6/Leu-6; T-cell surface glycoprotein CD1a | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
909 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
HI149 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P06126 | |
CD1A | |
Human thymocytes. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |