Learn More
CD19 Mouse anti-Human, APC-Cy7, Clone: SJ25C1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 557791
2064.03 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The SJ25C1 monoclonal antibody specifically binds to CD19, a B lymphocyte-lineage differentiation antigen. CD19, a 90-kDa transmembrance glycoprotein, is a member of the immunoglobulin superfamily and is expressed throughout B-lymphocyte development from the pro-B cell through the mature B-cell stages. Terminally differentiated plasma cells do not express CD19. On the surface of mature B cells, the CD19 molecule associates with CD21 (CR-2) and CD81 (TAPA-1), and this multimolecular complex synergizes with surface immunoglobulin to promote cellular activation. Studies with CD19-deficient mice have suggested that the level of CD19 expression affects the generation and maturation of B cells in the bone marrow and periphery. B-1 lineage B cells, also known as CD5+ B cells, are drastically reduced or absent in CD19-deficient mice. Increased levels of CD19 expression correlate with increased frequencies of peritonal and splenic B-1 cells and reduced numbers of conventional B lymphocytes in the periphery. CD19 participates in B-lymphocyte development, B-cell activation, maturation of memory B cells and regulation of tolerance. CD19 has also been detected on peritoneal mast cells, co-localized with CD21/CD35, and it is proposed to play a role in complement-mediated mast-cell activation.

Specifications
CD19 | |
Monoclonal | |
APC-Cyanine7 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
SJ25C1 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human NALM1 + NALM16 cells | |
100 Tests | |
Immunology | |
Human | |
IgG1 Îş |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.