Learn More
Thrombomodulin Monoclonal Antibody (141C01 (1009)), Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA511454
Additional Details : Weight : 0.01000kg
Description
MA5-11454 targets CD141 in IHC (P), FACS and ICC/IF applications and shows reactivity with Human and Rat samples. The MA5-11454 immunogen is recombinant protein encoding the six repeated EGF domains of human thrombomodulin.
Thrombomodulin Monoclonal antibody specifically detects Thrombomodulin in Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
Thrombomodulin | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 0.09% sodium azide | |
P07204 | |
THBD | |
Recombinant protein encoding the six repeated EGF domains of human thrombomodulin | |
RUO | |
7056, 83580 | |
4° C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
141C01 (1009) | |
Unconjugated | |
THBD | |
AHUS6; AI385582; BDCA3; BDCA-3; CD141; CD141 antigen; DC141; fetomodulin; sCD141; snoRNA MBII-339; soluble CD141; THBD; THPH12; THRM; thrombomdulin; thrombomodulin; thrombomodulin precursor; TM | |
Mouse | |
500 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 κ |