Få mere at vide
CD14 Mouse anti-Human, APC-Cy7, Clone: MφP9, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561709
Beschreibung
Reacts with a 53-55 kDa glycosylphosphatidylinositol (GPI)-anchored and single chain glycoprotein expressed at high levels on monocytes. Additionally, CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB. This antibody is suitable for staining acetone-fixed, frozen tissue sections.
Spezifikation
| CD14 | |
| Monoclonal | |
| APC-Cyanine7 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2b κ |
| Flow Cytometry | |
| MφP9 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human Monocytes | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.