Learn More
OX40 Polyclonal Antibody, PE, Invitrogen™
Goat Polyclonal Antibody
Brand: Thermo Scientific PA546881
Additional Details : Weight : 0.00010kg
Description
Less than 2% cross-reactivity with recombinant mouse (rm) EDAR, rm4-BB, rmCD27, rmDR3, rmGITR, rmNGF R, rmCD30, rmCD40, rmFas, rmOPG, rmRANK, rmTNF RI, and rmTNF RII is observed.
OX40 Polyclonal antibody specifically detects OX40 in Mouse samples. It is validated for Flow Cytometry.Specifications
OX40 | |
Polyclonal | |
Buffered saline solution with BSA and 0.09% sodium azide | |
P47741 | |
Tnfrsf4 | |
Mouse myeloma cell line NS0-derived recombinant mouse OX40/TNFRSF4 Val20-Pro211 | |
100 Tests | |
Primary | |
Mouse | |
Antibody | |
IgG |
Flow Cytometry | |
PE | |
Tnfrsf4 | |
ACT35; ACT35 antigen; ATC35 antigen; CD134; CD134 antigen; IMD16; Ly-70; lymphoid activation antigene ACT35; MRC OX40; Ox40; OX40 antigen; OX40 cell surface antigen; OX40 homologue; OX40L receptor; TAX transcriptionally-activated glycoprotein 1 receptor; tax-transcriptionally activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 receptor; TNF receptor superfamily member 4; Tnfrsf4; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor receptor superfamily member 4; tumor necrosis factor receptor superfamily, member 4; tumor necrosis factor superfamily, member 4; Txgp1; Txgp1l | |
Goat | |
Antigen affinity chromatography | |
RUO | |
22163 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |