missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GP130 Monoclonal Antibody (745314), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524355
Additional Details : Weight : 0.00010kg
Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
GP130 Monoclonal antibody specifically detects GP130 in Rat samples. It is validated for Flow Cytometry, Western Blot.Specifications
GP130 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P40190 | |
IL6ST | |
Mouse myeloma cell line NS0-derived recombinant rat gp130 Gln23-Glu618 (predicted) | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
745314 | |
Unconjugated | |
IL6ST | |
5133400A03Rik; AA389424; Ac1055; BB405851; CD130; CD130 antigen; CDW130; D13Ertd699e; EGK_16499; Gp130; gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form; gp130, oncostatin M receptor; GP130-RAPS; IL-6; IL-6 receptor subunit beta; IL-6R subunit beta; IL-6RB; IL-6R-beta; IL6R-beta; IL6ST; interleukin 6 signal transducer; interleukin 6 signal transducer (gp130, oncostatin M receptor); interleukin 6 signal transducer receptor; interleukin receptor beta chain; interleukin-6 receptor subunit beta; Interleukin-6 signal transducer; interleukin-6 signal transducer receptor; membrane glycoprotein; membrane glycoprotein 130; membrane glycoprotein gp130; oncostatin-M receptor subunit alpha | |
Mouse | |
Protein A/G | |
RUO | |
25205 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |