missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFR1 Monoclonal Antibody (HM104), PE, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA517899
Additional Details : Weight : 0.01000kg
Description
MA5-17899 targets CD120a/TNF Receptor I in FACS applications and shows reactivity with Mouse samples. The MA5-17899 immunogen is mouse TNFR-p55.
TNFR1 Monoclonal antibody specifically detects TNFR1 in Mouse samples. It is validated for Flow Cytometry.Specifications
TNFR1 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P25118 | |
Tnfrsf1a | |
Mouse TNFR-p55 | |
50 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry | |
HM104 | |
PE | |
Tnfrsf1a | |
CD120a; CD120a antigen; FPF; hypothetical protein; metalloproteinase inhibitor 4; MGC19588; MS5; n-smase activation assoc. factor; p55; p55 TNF receptor; p55-R; p60; p62; p63; sCD120a; soluble CD120a; soluble TNFR1; sTNF RI; sTNFR I; sTNFR1; sTNFRI; TBP1; TBPI; TIMP metallopeptidase inhibitor 4; Timp4; TIMP-4; tissue inhibitor of metalloproteinase 4; tissue inhibitor of metalloproteinases 4; TNF receptor alpha chain; TNF receptor superfamily member 1A; TNF-alphaR1; TNF-alpha-R1; TNFalpha-R1; TNFAR; TNFR; TNF-R; TNFR I; TNFR1; TNF-R1; Tnfr-1; TNFR1-d2; Tnfr-2; TNFR55; TNF-R55; TNFR60; TNFRI; TNF-RI; TNF-R-I; TNFR-I; TNFRp55; Tnfrsf1a; tumor necrosis factor binding protein 1; Tumor necrosis factor receptor; tumor necrosis factor receptor 1; tumor necrosis factor receptor 1A isoform beta; tumor necrosis factor receptor superfamily member 1A; Tumor necrosis factor receptor superfamily member 1A, membrane form; tumor necrosis factor receptor superfamily, member 1A; tumor necrosis factor rec | |
Rat | |
Protein G | |
RUO | |
21937 | |
4° C, do not freeze | |
Liquid |