missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calreticulin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA525922
Additional Details : Weight : 0.01000kg
Description
Calreticulin Polyclonal antibody specifically detects Calreticulin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Calreticulin | |
Polyclonal | |
Unconjugated | |
CALR | |
Autoantigen RO; CABP3; CALBP; calcium-binding protein 3; CALR; calrectulin; calregulin; Calreticulin; calreticulin precursor; cC1qR; CRP55; CRT; CRTC; Endoplasmic reticulum resident protein 60; epididymis secretory sperm binding protein Li 99n; ERp60; FLJ26680; grp60; HACBP; HEL-S-99n; I79_008346; RO; Ro/SS-A Antigen; Sicca syndrome antigen A (autoantigen Ro; calreticulin); SSA | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
12317, 64202, 811 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P14211, P18418, P27797 | |
CALR | |
recombinant protein of human CALR. | |
400 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |