missing translation for 'onlineSavingsMsg'
Få mere at vide

Calnexin Mouse, Unlabeled, Clone: 37, BD

Artikelnummer. 15885499
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
15885499 50 μg
15895499 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15885499 Leverandør BD Biosciences Leverandørnr. 610523

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody

Calnexin is a 90kDa integral membrane protein located primarily in the ER. It contains a long N-terminal calcium-binding domain that extends into the lumen of the ER and a short, acidic cytosolic domain. Calnexin associates with several cell surface proteins as they pass through the ER. Since it also forms complexes with other resident ER proteins involved in the Ca2+-dependent retention of proteins, calnexin may act as a chaperone. The amino acid sequence of calnexin is highly conserved among species and is similar in sequence to calreticulin, another Ca2+-binding protein found in the ER. Neither calnexin nor calreticulin contain the calcium-binding motif, known as the “E-F hand” found in the calmodulin family of proteins.

Host Species: Mouse
Clone: 37
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human Calnein aa. 116-301

Immunofluorescence, Immunoprecipitation

TRUSTED_SUSTAINABILITY

Tekniske data

Antigen Calnexin
Applikationer Western Blot
Klassifikation Monoclonal
Klon 37
Koncentration 250μg/mL
Konjugeret Unconjugated
Formulering Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Værtsarter Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Oprensningsmetode Affinity Purified
Mængde 50 μg
Regulatorisk status RUO
Forskningsdisciplin Cell Biology
Primær eller sekundær Primary
Målarter Canine, Human, Mouse, Rat
Indhold og opbevaring Store undiluted at -20°C.
Isotype IgG1
Vis mere Vis mindre

For Research Use Only.

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.