missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
beta Tubulin Loading Control, Biotin, clone: BT7R, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516308BTIN
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
MA5-16308-BTIN has successfully been used for Western blot, IHC (P), FACS, ELISA and ICC/IF
Beta tubulins are one of two core protein families (alpha and beta tubulins) that heterodimerize and assemble to form microtubules. Beta-III tubulin is primarily expressed in neurons and may be involved in neurogenesis, axon guidance and maintece. Mutations in the beta tubulin gene are the cause of congenital fibrosis of the extraocular muscles type 3. Beta-III tubulin was also detected in Sertoli cells of the testis and transiently in non-neuronal embryonic tissues. Glutamate residues at the C-terminus of beta III tubulin can be glutamylated. The precise function of such modifications is unclear. Tubulin is phosphorylated on Ser-172 by CDK1 during cell cycle progression. Ser-172 phosphorylation inhibits tubulin incorporation into microtubules. Microtubules, the major cytoskeletal elements found in all eukaryotic cells, consist of Tubulin, which is a dimer of two 55 kDa subunits: alpha and Beta. Microtubules play key roles in chromosome segregation in mitosis, intracellular transport, ciliary and flagellar bending, and structural support of the cytoskeleton. Because beta-tubulin is ubiquitously expressed in all eukaryotic cells, it is frequently used as a loading control for assays involving protein detection, such as Western blotting.
Tekniske data
| Beta-Tubulin Loading Control | |
| Monoclonal | |
| 1mg/mL | |
| TUBB | |
| beta 5-tubulin, beta Ib tubulin, beta-4 tubulin, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, tubulin beta-5 chain, tubulin, beta polypeptide | |
| Mouse | |
| Protein A | |
| RUO | |
| 100347858, 100358689, 203068, 396254, 396427, 545486, 679312, 81027 | |
| Store at 4°C. Do not freeze. | |
| Liquid |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| BT7R | |
| Biotin | |
| P07437, Q9H4B7, A2AQ07 | |
| TUBB, TUBB1 | |
| KLH conjugated peptide of the N-terminus of Beta-Tubulin. | |
| 50 μL | |
| Primary | |
| Chicken, Human, Mouse, Non-human Primate, Rabbit, Rat | |
| Antibody | |
| IgG2a |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion