missing translation for 'onlineSavingsMsg'
Learn More

Intestinal Alkaline Phosphatase Monoclonal Antibody (P4A10E4), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA514742

Additional Details : Weight : 0.01000kg

Product Code. 11315203

  • 3870.00 DKK / 100µg
Estimated Shipment: 08-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MA5-14742 targets Alkaline Phosphatase in ELISA, IHC, IP, and RIA applications and shows reactivity with Bovine samples. The MA5-14742 immunogen is purified Bovine Calf Intestine ALP. MA5-14742 detects Alkaline Phosphatase which has a predicted molecular weight of approximately 57 kDa. Product MA514742 is a smaller package size of MIA1703 (formerly sold as a Seradyn product).

Intestinal Alkaline Phosphatase Monoclonal antibody specifically detects Intestinal Alkaline Phosphatase in Bovine samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA).
Specifications

Specifications

Intestinal Alkaline Phosphatase
Monoclonal
1 mg/mL
0.01M potassium phosphate with 0.135M NaCl, 1mg/mL BSA, 2.5mM KCl and 0.05% sodium azide
P19111
ALPI
Purified Bovine Calf Intestine ALP
100 μg
Primary
Bovine
Antibody
IgG1
ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA)
P4A10E4
Unconjugated
ALPI
2010001C14Rik; Akp2; Akp6; Alkaline phosphatase 1, intestinal, defined by SSR; alkaline phosphatase 6; alkaline phosphatase, intestinal; alkaline phosphomonoesterase; Alp1; ALPI; AP; EC 3.1.3.1; gIAP; glycerophosphatase; I Kasahara isozyme; IAP; IAP-1; IAP-I; intestinal alkaline phosphatase; intestinal alkaline phosphatase 1; intestinal alkaline phosphatase I IAP-I; intestinal alkaline phosphatase III; intestinal alkaline phosphatase IV; intestinal alkaline phosphatase V; intestinal-type alkaline phosphatase; intestinal-type alkaline phosphatase 1; Kasahara isozyme; PLAP-1; Regan isozyme; S51097
Mouse
Ion-exchange chromatography
RUO
280993
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers