missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ ALDH1B1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528554
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Recommended positive controls: A431. Predicted reactivity: Mouse (94%), Rat (94%), Chimpanzee (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Aldehyde dehydrogenases (ALDHs) mediate NADP+-dependent oxidation of aldehydes into acids during detoxification of alcohol-derived acetaldehyde, lipid peroxidation and metabolism of corticosteroids, biogenic amines and neurotransmitters. Alcohol drinking habits and cardiovascular disease risk factors may be associated with ALDH gene variants. ALDH1B1 (Aldehyde dehydrogenase family 1 member B1), also known as ALDH5 or ALDHX (Aldehyde dehydrogenase X, mitochondrial), is a 517 amino acid mitochondrial protein that is expressed in the liver, testis and to a lesser extent in brain. ALDH1B1 belongs to the aldehyde dehydrogenase family and may play a major role in ethanol detoxification.
Tekniske data
| ALDH1B1 | |
| Polyclonal | |
| Unconjugated | |
| ALDH1B1 | |
| 2700007F14Rik; acetaldehyde dehydrogenase 5; aldehyde dehydrogenase 1 family member B1; aldehyde dehydrogenase 1 family, member B1; aldehyde dehydrogenase 1B1; aldehyde dehydrogenase 5; Aldehyde dehydrogenase family 1 member B1; aldehyde dehydrogenase X, mitochondrial; ALDH class 2; ALDH1B1; ALDH5; ALDHX; MGC2230 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 219, 72535 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.87 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P30837, Q9CZS1 | |
| ALDH1B1 | |
| Recombinant protein encompassing a sequence within the center region of human ALDH1B1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion