missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
anti-AKR1D1 Matched Antibody Pair, Abnova
Brand: Abnova H00006718-AP21.1Set
Indeholder: Rabbit MaxPab™ affinity purified polyclonal anti-AKR1D1 (100μg); Mouse polyclonal anti-AKR1D1 (40μL); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Beskrivelse
The enzyme encoded by this gene is responsible for the catalysis of the 5-β-reduction of bile acid intermediates and steroid hormones carrying a δ(4)-3-one structure. Deficiency of this enzyme may contribute to hepatic dysfunction.
Host Species: Rabbit; Mouse
Species Reactivity: Human
ELISA
Tekniske data
| AKR1D1 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human AKR1D1. | |
| 3o5bred/SRD5B1 | |
| Mouse, Rabbit | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-AKR1D1 (100 μg) 2. Detection antibody: mouse polyclonal anti-AKR1D1 (40 ul) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| AKR1D1 | |
| AKR1D1 | |
| Affinity Purified | |
| RUO | |
| 6718 | |
| Human | |
| Serum |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion