missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AF9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA513174
Additional Details : Weight : 0.01000kg
Description
AF9 Polyclonal antibody specifically detects AF9 in Human, Mouse samples. It is validated for Western Blot.Specifications
AF9 | |
Polyclonal | |
Unconjugated | |
MLLT3 | |
2210011H10Rik; 2610012I03Rik; 3830408D16Rik; Af9; Af-9; AF-9 protein; ALL1-fused gene from chromosome 9 protein; D4Ertd321e; MLLT3; MLLT3, super elongation complex subunit; myeloid/lymphoid or mixed lineage-leukemia translocation to 3 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocated to, 3; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 3; myeloid/lymphoid or mixed-lineage leukemia translocated to chromosome 3 protein; Myeloid/lymphoid or mixed-lineage leukemia translocated to chromosome 3 protein homolog; myeloid/lymphoid or mixed-lineage leukemia, translocated to, 3; myeloid/lymphoid or mixed-lineage leukemia,translocated to, 3 (trithorax homolog, Drosophila); myeloid/lymphoid or mixed-lineage leukemia; translocated to 3; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 3; protein AF-9; translocated to, 3; YEATS domain-containing protein 3; YEATS3 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
4300, 70122 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
A2AM29, P42568 | |
MLLT3 | |
KLH conjugated synthetic peptide between 407-438 amino acids from the central region of human AF9 (MLLT3) | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |