missing translation for 'onlineSavingsMsg'
Learn More
Learn More
membrane-associated ring finger (C3HC4) 6, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MARCH6.
Brand: Abnova H00010299-A01.50uL
Additional Details : Weight : 0.00010kg
Description
Sequence: DTAEEDICRVCRSEGTPEKPLYHPCVCTGSIKFIHQECLVQWLKHSRKEYCELCKHRFAFTPIYSPDMPSRLPIQDIFAGLVTSIGTAIRSpecifications
membrane-associated ring finger (C3HC4) 6 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant MARCH6. | |
MARCH6 | |
KIAA0597/MARCH-VI/RNF176/TEB4 | |
Mouse | |
50 μL | |
Ubiquitin/Proteasome | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_005885 | |
MARCH6 | |
40974 (NP_005876, 2 a.a. ∼ 91 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
10299 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |