Learn More
Anthraquinone, TRC
Product Code. 30134951
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134951 500 g 500g
30136801 250 g 250g
17939339 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 30134951 Supplier Toronto Research Chemicals Supplier No. TRCA679245500G
 Restricted Item
Forventet forsendelse: 03-12-2026
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 84-65-1
Molecular Formula C14 H8 O2
Molecular Weight (g/mol) 208.21
Synonym Anthraquinone (8CI), 1,4,11,12-Tetrahydro-9,10-anthraquinone, 9,10-Anthraquinone, Anthracene-9,10-quinone, Anthradione, Avipel, DAQ-N, Flight Control, Flight Control Plus, Hoelite, Kawasaki SAQ, Morkit, NSC 7957, SAQ, SAQ (quinone)
IUPAC Name anthracene-9,10-dione
SMILES O=C1c2ccccc2C(=O)c3ccccc13
Specifications
Quantity 500 g
InChI Formula InChI=1S/C14H8O2/c15-13-9-5-1-2-6-10(9)14(16)12-8-4-3-7-11(12)13/h1-8H
Formula Weight 208.0524
Chemical Name or Material Anthraquinone