Få mere at vide
Anthranilic-3,4,5,6-d4 Acid, CDN
Artikelnummer. 30320662
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320662 0.1 g 0.1g
30313497 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320662 Leverandør CDN Leverandørnr. CDND43940.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Anthranilic-3,4,5,6-d4 acid
CAS 60124-83-6
Molekylær formel C7 2H4 H3 N O2
Mængde 0.1 g
Synonym Benzoic-2,3,4,5-d4 acid, 6-amino-, 6-Aminobenzoic-2,3,4,5-d4 acid, 2-Aminobenzoic-3,4,5,6-d4 acid, Anthranilic acid-d4, Anthranilic acid-d4 (phenyl-d4), Anthranilic-3,4,5,6-D4 acid
InChI formel InChI=1 S/C7H7NO2/c8-6-4-2-1-3-5(6)7(9)10/h1-4 H,8H2,(H,9,10)/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=O)O)c(N)c1[2 H]
IUPAC navn 2-amino-3,4,5,6-tetradeuteriobenzoic acid
Molekylvægt (g/mol) 141.16
Formel vægt 141.0728 g/mol
Procent renhed 97 atom % D, min 98% Chemical Purity