Få mere at vide
Antazoline Phosphate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Antazoline Phosphate |
| CAS | 154-68-7 |
| Molekylær formel | C17 H19 N3 . H3 O4 P |
| Mængde | 10 g |
| Synonym | Antazoline Phosphate, 4,5-Dihydro-N-phenyl-N-(phenylmethyl)-1H-imidazole-2-methanamine phosphate (1:1), 2-[(N-Benzylanilino)methyl]-2-imidazoline phosphate (1:1), 2-[(N-Benzylanilino)methyl]-2-imidazoline dihydrogen phosphate, Antazoline phosphate, Antistine phosphate |
| InChI formel | InChI=1S/C17H19N3.H3O4P/c1-3-7-15(8-4-1)13-20(14-17-18-11-12-19-17)16-9-5-2-6-10-16;1-5(2,3)4/h1-10H,11-14H2,(H,18,19);(H3,1,2,3,4) |
| SMIL | OP(=O)(O)O.C(N(Cc1ccccc1)c2ccccc2)C3=NCCN3 |
| IUPAC navn | N-benzyl-N-(4,5-dihydro-1H-imidazol-2-ylmethyl)aniline;phosphoric acid |
| Molekylvægt (g/mol) | 363.35 |
| Formel vægt | 363.1348 |