Få mere at vide
Tocris Bioscience™ ANR 94
GREENER_CHOICE
Artikelnummer. 18014782
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18014782 10 mg
18083872 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18014782 Leverandør Tocris Bioscience™ Leverandørnr. 3937/10
Adenosine A2A antagonist
Adenosine A2A receptor (AA2AR) antagonist (Ki values are 643 and 46 nM for rat and human AA2ARs respectively). Most active AA2AR antagonist for human receptors. Displays activity in the treatment of Parkinson's disease in vivo; improves parkinsonian motor deficits and tremors. Exhibits neuroprotective and anti-inflammatory effects.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 634924-89-3
Molekylær formel C9H13N5O
Tekniske data
Kemisk navn eller materiale ANR 94
Mængde 10 mg
Formel vægt 207.23
Renhed >99%
Molekylvægt (g/mol) 207.237