Description
Annexin A6 Polyclonal specifically detects Annexin A6 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Conjugate | FITC |
| Dilution | Western Blot |
| Formulation | PBS |
| Regulatory Status | RUO |
| Research Discipline | Endosome Markers |
| Content And Storage | Store at 4C in the dark. |
| Product Type | Antibody |
| Product Line | Novus |