Learn More
Anisole, 99%, Thermo Scientific Chemicals
Catalog Number p-7020890
Click to view available options
Quantity:
500 g
2500 g
100-66-3
C7H8O
108.14
MFCD00008354
RDOXTESZEPMUJZ-UHFFFAOYSA-N
methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene
7519
CHEBI:16579
anisole
COC1=CC=CC=C1
100-66-3
C7H8O
108.14
MFCD00008354
RDOXTESZEPMUJZ-UHFFFAOYSA-N
methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene
7519
CHEBI:16579
anisole
COC1=CC=CC=C1
Anisole is widely used as a solvent for the synthesis of various organic compounds, anethole, nonylphenol isomer 4-(3',6'-dimethyl-3-heptyl)phenol, perfumes, insect pheromones and pharmaceuticals. It finds application in the preparation of inorganic complexes and materials such as tin-core/tin oxide nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Anisole is widely used as a solvent for the synthesis of various organic compounds, anethole, nonylphenol isomer 4-(3′,6′-dimethyl-3-heptyl)phenol, perfumes, insect pheromones and pharmaceuticals. It finds application in the preparation of inorganic complexes and materials such as tin-core/tin oxide nanoparticles.
Solubility
Miscible with methanol and acetone. Immiscible with water.
Notes
Keep away from sources of ignition. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -37°C
Density 0.994
Boiling Point 153°C to 154°C
Flash Point 43°C (109°F)
Odor Ether-like
Refractive Index 1.517
Quantity 500 g
UN Number UN2222
Beilstein 506892
Merck Index 14,669
Solubility Information Miscible with methanol and acetone. Immiscible with water.
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material Anisole