Få mere at vide
Anisole-2,3,4,5,6-d5, CDN
Artikelnummer. 30320537
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320537 1 g -
30318884 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320537 Leverandør CDN Leverandørnr. CDND10391G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Anisole-2,3,4,5,6-D5
CAS 50629-14-6
Molekylær formel C7 D5 H3 O
Mængde 1 g
Synonym Benzene-d5, methoxy- (9 CI, ACI), Anisole-2,3,4,5,6-d5 (6 CI), Methoxybenzene-d5 (ACI), 1,2,3,4,5-Pentadeuterio-6-methoxybenzene, 2,3,4,5,6-Pentadeuterioanisole, Anisole-2,3,4,5,6-D5, Anisole-d5
InChI formel InChI=1 S/C7H8O/c1-8-7-5-3-2-4-6-7/h2-6 H,1H3/i2D,3 D,4 D,5 D,6 D
SMIL [2 H]c1c([2 H])c([2 H])c(OC)c([2 H])c1[2 H]
IUPAC navn 1,2,3,4,5-pentadeuterio-6-methoxybenzene
Molekylvægt (g/mol) 113.1686
Formel vægt 113.0889 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity