Learn More
STEMCELL Technologies Animal Component-Free Cell Dissociation Kit
Description
- Animal Component-Free Cell Dissociation Kit is optimized for dissociation and passaging of human stem and progenitor cells cultured in various media formulations
- Animal Component-Free Cell Dissociation Kit includes ACF Enzymatic Dissociation Solution and ACF Enzyme Inhibition Solution
- This kit was previously named MesenCult™-ACF Dissociation Kit
- The name was changed to accommodate the use of this product with other cell types
Specifications
Specifications
| Cell Type | Endothelial Cells, Endothelial Colony Forming Cells (ECFCs), HUVECs, Mesenchymal Cells, PSC-Derived, Mesenchymal Stem and Progenitor Cells, Myogenic Stem and Progenitor Cells |
| Quantity | 1 Kit |
Product Title
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?