Få mere at vide
Aniline-3,5-d2, CDN
Artikelnummer. 30314033
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30314033 0.25 g -
30310904 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314033 Leverandør CDN Leverandørnr. CDND81430.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Aniline-3,5-D2
CAS 19617-82-4
Molekylær formel C6 D2 H5 N
Mængde 0.25 g
Synonym Benzen-3,5-d2-amine (9 CI), Aniline-3,5-d2 (7 CI, 8 CI), 3,5-Dideuterioaniline, 3,5-Dideuteroaniline, Aniline-3,5-D2
InChI formel InChI=1 S/C6H7N/c7-6-4-2-1-3-5-6/h1-5 H,7H2/i2D,3 D
SMIL [2 H]c1cc([2 H])cc(N)c1
IUPAC navn 3,5-dideuterioaniline
Molekylvægt (g/mol) 95.1388
Formel vægt 95.0704 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity