Få mere at vide
Anhydrodihydroartemisinin, Mikromol™
Artikelnummer. 30049525
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049525 25 mg
17914076 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049525 Leverandør Mikromol Leverandørnr. MM3253.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Anhydrodihydroartemisinin
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 9,10-Anhydrodehydroartemisinin
CAS 82596-30-3
Analyt- eller komponentnavne 9,10-Anhydrodehydroartemisinin
Forsendelsestilstand Room Temperature
Molekylær formel C15 H22 O4
Certificeringer/compliance ISO 17025
Synonym (3R,5aS,6R,8aS,12R,12aR)-3,4,5,5a,6,7,8,8a-Octahydro-3,6,9-trimethyl--3,12-epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin, (+)-9,10-Anhydrodehydroartemisinin, 9,10-Anhydro-10-deoxoartemisinin, 9,10-Anhydrodehydroartemisinin, Anhydrodihydroartemisinin, Anhydrodihydroartemisnin, 3, 2-Epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin, 3,4,5,5a,6,7,8,8a-octahydro-3,6,9-trimethyl-, [3R-(3alpha,5abeta,6beta,8abeta,12beta,12aR*)]-, Artesunate Related Compound C
InChI formel InChI=1S/C15H22O4/c1-9-4-5-11-10(2)8-16-13-15(11)12(9)6-7-14(3,17-13)18-19-15/h8-9,11-13H,4-7H2,1-3H3/t9-,11+,12+,13-,14-,15+/m1/s1
SMIL C[C@@H]1CC[C@H]2C(=CO[C@@H]3O[C@@]4(C)CC[C@@H]1[C@@]23OO4)C
IUPAC navn (3R,5aS,6R,8aS,12R,12aR)-3,4,5,5a,6,7,8,8a-Octahydro-3,6,9-trimethyl--3,12-epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin
Molekylvægt (g/mol) 266.33
Mængde 25 mg
Formel vægt 266.1518 g/mol