missing translation for 'onlineSavingsMsg'
Få mere at vide

Angiotensin (1-7) xTFA salt, TRC

Artikelnummer. 30329834
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30329834 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30329834 Leverandør Toronto Research Chemicals Leverandørnr. TRCA63794550MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Angiotensin (1-7) xTFA salt
CAS 51833-78-4
Molekylær formel C41H62N12O11 . xTFA
Mængde 50 mg
Synonym 5-L-Isoleucine-1-7-angiotensin II, 1-7-Human angiotensin, 1-7-Human angiotensin 1, 1-7-Human angiotensin 2, 1-7-Human angiotensin I, 1-7-Human angiotensin II, 1-7-[Ile5]-angiotensin, 115: PN: WO2006032674 SEQID: 115 claimed sequence, 11: PN: WO0212471 SEQID: 18 unclaimed sequence, 11: PN: WO2010028845 SEQID: 9 claimed sequence, 18: PN: US6610497 SEQID: 18 unclaimed sequence, 18: PN: US6884771 SEQID: 18 unclaimed sequence, 18: PN: US6989363 SEQID: 18 unclaimed sequence, 1: PN: US20140205631 SEQID: 4 claimed sequence, 1: PN: US20140296143 SEQID: 1 claimed protein, 1: PN: US20160016996 SEQID: 2 claimed protein, 1: PN: US8633158 SEQID: 1 claimed protein, 1: PN: WO2010028845 SEQID: 1 claimed protein, 1: PN: WO2012021578 SEQID: 1 claimed protein, 1: PN: WO2013032784 SEQID: 1 claimed protein, 1: PN: WO2013090833 SEQID: 1 claimed sequence, 1: PN: WO2013119870 SEQID: 1 claimed sequence, 1: PN: WO2013158959 SEQID: 1 claimed protein, 1: PN: WO2014190152 SEQID: 1 claimed protein, 1: PN: WO2015054005 SEQID: 1 claimed protein, 1: PN: WO2016112247 SEQID: 1 claimed protein, 1: PN: WO2017066552 SEQID: 1 claimed protein, 28: PN: US20160160263 SEQID: 67 claimed protein, 2: PN: WO2014021942 SEQID: 4 claimed protein, 39: PN: US20090227507 SEQID: 1 unclaimed protein, 3: PN: WO0144270 SEQID: 4 claimed protein, 3: PN: WO2008089532 PAGE: 19 unclaimed sequence, 3: PN: WO2011053789 SEQID: 3 claimed protein, 3: PN: WO2013158628 SEQID: 3 unclaimed protein, 3: PN: WO2013172956 SEQID: 4 unclaimed protein, 3: PN: WO2013182237 SEQID: 3 claimed protein, 453: PN: WO0069900 SEQID: 639 unclaimed sequence, 4: PN: WO0056345 SEQID: 4 claimed sequence, 4: PN: WO0143761 SEQID: 4 claimed protein, 4: PN: WO0155176 SEQID: 4 claimed protein, 4: PN: WO0198325 SEQID: 4 claimed protein, 4: PN: WO02087504 SEQID: 4 claimed protein, 4: PN: WO03039434 PAGE: 7 unclaimed sequence, 5-Ile-angiotensin-(1-7), 529: PN: US20090175821 SEQID: 639 claimed protein, 5: PN: US20030130196 SEQID: 4 claimed protein, 6: PN: WO0018899 PAGE: 18 unclaimed sequence, 8: PN: WO03072059 SEQID: 1 claimed protein, Human angiotensin 1-(1-7), Human angiotensin 2-(1-7), Human angiotensin I-(1-7), Human angiotensin II-(1-7), Human angiotensin-(1-7), TXA 127, [Ile5]angiotensin I (1-7)
InChI formel InChI=1S/C41H62N12O11/c1-5-22(4)33(38(61)50-29(17-24-19-45-20-47-24)39(62)53-15-7-9-30(53)40(63)64)52-36(59)28(16-23-10-12-25(54)13-11-23)49-37(60)32(21(2)3)51-35(58)27(8-6-14-46-41(43)44)48-34(57)26(42)18-31(55)56/h10-13,19-22,26-30,32-33,54H,5-9,14-18,42H2,1-4H3,(H,45,47)(H,48,57)(H,49,60)(H,50,61)(H,51,58)(H,52,59)(H,55,56)(H,63,64)(H4,43,44,46)/t22-,26-,27-,28-,29-,30-,32-,33-/m0/s1
SMIL CC[C@H](C)[C@H](NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@@H](NC(=O)[C@H](CCCNC(=N)N)NC(=O)[C@@H](N)CC(=O)O)C(C)C)C(=O)N[C@@H](Cc2cnc[nH]2)C(=O)N3CCC[C@H]3C(=O)O
IUPAC navn (2S)-1-[(2S)-2-[[(2S,3S)-2-[[(2S)-2-[[(2S)-2-[[(2S)-2-[[(2S)-2-amino-3-carboxypropanoyl]amino]-5-(diaminomethylideneamino)pentanoyl]amino]-3-methylbutanoyl]amino]-3-(4-hydroxyphenyl)propanoyl]amino]-3-methylpentanoyl]amino]-3-(1H-imidazol-5-yl)propanoyl]pyrrolidine-2-carboxylic acid
Molekylvægt (g/mol) 899
Formel vægt 898.4661
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.