Learn More
Andropanolide, MedChemExpress
MedChemExpress Andrographolide (Andro) is a small antagonist for NF-κB activation by covalent modifying reduced cysteine 62 of p50.

1873.25 DKK - 4683.12 DKK
Specifications
Chemical Name or Material | Andropanolide |
---|---|
CAS | 869807-57-8 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C20H30O5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16431658
![]() |
Medchem Express
HY-N1912 |
5 mg |
4683.12 DKK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16422579
![]() |
Medchem Express
HY-N1912 |
1 mg |
1873.25 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Andrographolide is a bicyclic diterpenoid lactone mainly produced from the plant Andrographis (Andrographis paniculate). Andrographolide suppresses the activation of NF-κB in stimulated endothelial cells, which reduces the expression of cell adhesion molecule E-selectin and prevents E-selectin-mediated leukocyte adhesion, but has no effect on IκBα degradation, p50 and p65 nuclear translocation.Specifications
Andropanolide | |
White | |
C20H30O5 | |
350.45 | |
98.78% | |
Powder |
869807-57-8 | |
Research | |
O=C1OC[C@@H](O)/C1=C/C[C@@H]2C(CC[C@]3([H])[C@](C)(CO)[C@H](O)CC[C@@]23C)=C | |
350.45 | |
Research | |
COVID-19-immunoregulation |
Safety and Handling
missing translation for 'shelfLife' : 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Recommended Storage : 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.