Få mere at vide
(+)-Anatoxin A Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (+)-Anatoxin A Hydrochloride |
| CAS | 64314-16-5 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C10 H15 N O . Cl H |
| Mængde | 2.5 mg |
| Synonym | Ethanone, 1-(1R,6R)-9-azabicyclo[4.2.1]non-2-en-2-yl-, hydrochloride (9CI), Ethanone, 1-(9-azabicyclo[4.2.1]non-2-en-2-yl)-, hydrochloride, (1R)-, 9-Azabicyclo[4.2.1]nonane, ethanone deriv., (+)-Anatoxin-a hydrochloride, Anatoxin a hydrochloride, Anatoxin a hydrochloride, (+)-Anatoxin A hydrochloride |
| InChI formel | InChI=1S/C10H15NO.ClH/c1-7(12)9-4-2-3-8-5-6-10(9)11-8;/h4,8,10-11H,2-3,5-6H2,1H3;1H/t8-,10-;/m1./s1 |
| SMIL | Cl.CC(=O)C1=CCC[C@@H]2CC[C@H]1N2 |
| IUPAC navn | 1-[(1R,6R)-9-azabicyclo[4.2.1]non-4-en-5-yl]ethanone;hydrochloride |
| Molekylvægt (g/mol) | 201.69 |
| Vis mere |