Learn More
Abeba™ Anatom 32120 Shoes
Product Code. 15495814
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Unit Size:
1 pair
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15495814 52 1 pair
15435764 36 1 pair
15455764 37 1 pair
15485764 38 1 pair
15495764 39 1 pair
15415774 40 1 pair
15455774 41 1 pair
15445784 42 1 pair
15485784 43 1 pair
15405794 44 1 pair
15425794 45 1 pair
15445794 46 1 pair
15475794 47 1 pair
15445804 48 1 pair
15495804 49 1 pair
15425814 50 1 pair
15465814 51 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Product Code. 15495814 Supplier Abeba™ Supplier No. 32120/52
Forventet forsendelse: 02-12-2026
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 SB A, E SRC
  • Microfiber
  • White
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3582)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Breathable lining with Silver ions
  • Instep area adjustable, with elastic band
  • velcro fastening
  • suitable for heel and foot lift
  • Suitable for hygienically demanding areas
Specifications
Certifications/Compliance CE EN ISO 20345:2011 SB A, E SRC
Color White
Material Microfiber
Size 52
Sole Features Slip-resistant
Sole Material Polyurethane
Style 32120
Toe Material Steel