missing translation for 'onlineSavingsMsg'
Få mere at vide

Anastrozole-d12, TRC

Artikelnummer. 30338283
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30338283 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30338283 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6374271MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Anastrozole-d12
CAS 120512-32-5
Renhedsgrad noter HPLC
Molekylær formel C17H7D12N5
Mængde 1 mg
Synonym α1,α1,α3,α3-(Tetramethyl-d12)-5-(1H-1,2,4-triazol-1-ylmethyl)-1,3-benzenediacetonitrile, 2-[3-(2-Cyano-2-propyl)-5-(1,2,4-triazol-1-ylmethyl)phenyl]-2 -methylpropiononitrile-d12, Anastrazole-d12, Anastrol-d12, Arimidex-d12, ICI-D 1033-d12, ZD 1033-d12
InChI formel InChI=1S/C17H19N5/c1-16(2,9-18)14-5-13(8-22-12-20-11-21-22)6-15(7-14)17(3,4)10-19/h5-7,11-12H,8H2,1-4H3/i1D3,2D3,3D3,4D3
SMIL [2H]C([2H])([2H])C(C#N)(C([2H])([2H])[2H])C1=CC(C(C#N)(C([2H])([2H])[2H])C([2H])([2H])[2H])=CC(CN2C=NC=N2)=C1
IUPAC navn 2-[3-(2-cyano-1,1,1,3,3,3-hexadeuteriopropan-2-yl)-5-(1,2,4-triazol-1-ylmethyl)phenyl]-3,3,3-trideuterio-2-(trideuteriomethyl)propanenitrile
Molekylvægt (g/mol) 305.44
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.