Få mere at vide
Anastrozole-d12, TRC
Artikelnummer. 30342768
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30342768 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30342768 Leverandør Toronto Research Chemicals Leverandørnr. TRCA63742710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Anastrozole-d12
CAS 120512-32-5
Renhedsgrad noter HPLC
Molekylær formel C17H7D12N5
Mængde 10 mg
Synonym α1,α1,α3,α3-(Tetramethyl-d12)-5-(1H-1,2,4-triazol-1-ylmethyl)-1,3-benzenediacetonitrile, 2-[3-(2-Cyano-2-propyl)-5-(1,2,4-triazol-1-ylmethyl)phenyl]-2 -methylpropiononitrile-d12, Anastrazole-d12, Anastrol-d12, Arimidex-d12, ICI-D 1033-d12, ZD 1033-d12
InChI formel InChI=1S/C17H19N5/c1-16(2,9-18)14-5-13(8-22-12-20-11-21-22)6-15(7-14)17(3,4)10-19/h5-7,11-12H,8H2,1-4H3/i1D3,2D3,3D3,4D3
SMIL [2H]C([2H])([2H])C(C#N)(C([2H])([2H])[2H])C1=CC(C(C#N)(C([2H])([2H])[2H])C([2H])([2H])[2H])=CC(CN2C=NC=N2)=C1
IUPAC navn 2-[3-(2-cyano-1,1,1,3,3,3-hexadeuteriopropan-2-yl)-5-(1,2,4-triazol-1-ylmethyl)phenyl]-3,3,3-trideuterio-2-(trideuteriomethyl)propanenitrile
Molekylvægt (g/mol) 305.44
Procent renhed >95