missing translation for 'onlineSavingsMsg'
Få mere at vide

Anabaseine, TRC

Artikelnummer. 30339431
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30339431 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30339431 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6371501G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Anabaseine
CAS 3471-05-4
Molekylær formel C10 H12 N2
Mængde 1 g
InChI formel InChI=1S/C10H12N2/c1-2-7-12-10(5-1)9-4-3-6-11-8-9/h3-4,6,8H,1-2,5,7H2
SMIL C1CCC(=NC1)c2cccnc2
IUPAC navn 3-(2,3,4,5-tetrahydropyridin-6-yl)pyridine
Molekylvægt (g/mol) 160.22
Formel vægt 160.1
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.