Få mere at vide
Tocris Bioscience™ AMZ 30
Beskrivelse
AMZ 30 is a potent, selective and irreversible inhibitor of protein phosphatase methylesterase-1 (PME-1; IC50 = 600 nM) with > 100-fold selectivity for PME-1 over other serine hydrolases in human cell lysates. AMZ 30 inactivates PME-1 and reduces the demethylated form of serine/threonine protein phosphatase 2A (PP2A) in HEK 293T cells. AMZ 30 perturbs leucine carboxyl methyltransferase-1 (LCMT1)-PME-1 methylation equilibrium leading to shortening of mitotic spindles and mitotic arrest in HeLa cells. It also shows antiproliferative properties in an in vitro model of endometrial carcinoma.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2 E)-2-[(4-Fluorophenyl)sulfonyl]-3-[1-[(3-nitrophenyl)sulfonyl]-1 H-pyrrol-2-yl]-2-propenenitrile |
| CAS | 1313613-09-0 |
| Mængde | 10 mg |
| Mål | Hydrolase Inhibitors |
| Molekylær formel | C19H12FN3O6S2 |
| Renhed | 0.98 |