missing translation for 'onlineSavingsMsg'
Learn More

Amylmetacresol, LoGiCal

Product Code. 30457849
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30457849 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457849 Supplier LoGiCal Supplier No. LGCFOR1132.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCals certificerede referencematerialer som smarte, native løsninger og stabile isotop-mærkede løsninger, med ekspertdokumentation og support. Fra retsmedicinske til kliniske laboratorier – vælg LoGiCal for selvtillid, overholdelse og analytisk ekspertise.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Amylmetacresol
CAS 1300-94-3
Analyt- eller komponentnavne Amylmetacresol
Forsendelsestilstand Stuetemperatur
Molekylær formel C12 H18 O
Certificeringer/compliance ISO 9001
Synonym Amylmetacresol, 5-Methyl-2-pentylphenol
InChI formel InChI=1S/C12H18O/c1-3-4-5-6-11-8-7-10(2)9-12(11)13/h7-9,13H,3-6H2,1-2H3
Løsningstype Fedt
SMIL CCCCCc1ccc(C)cc1O
IUPAC navn 5-methyl-2-pentylphenol
Industry Type Klinisk testning
Molekylvægt (g/mol) 178.27
Mængde 10 mg
Formel vægt 178,1358 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.