missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Amylin, Human, Amide

For cellular and molecular biology applications

2976.00 DKK - 4374.00 DKK

Specifications

Species Human
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 3860.16
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15468339
SDS View Documents Promotion Details
Thermo Scientific™
J66825.!D
0.5 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15478339
SDS View Documents Promotion Details
Thermo Scientific™
J66825.M^
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Human
-30°C to -10°C
3860.16
RUO - research use only
HPLC
Solid
Amylin, human, amide
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only