Få mere at vide
MedChemExpress Amylin, amide, human
Artikelnummer. 16442829
Encompass_Preferred
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
16442829 1 mg
1 options
Artikelnummer. 16442829 Leverandør MedChemExpress Leverandørnr. HYP1070
MedChemExpress Amylin, amide, human, a 37-amino acid polypeptide, is a pancreatic hormone cosecreted with insulin that exerts unique roles in metabolism and glucose homeostasis. Amylin, amide, human inhibits glucagon secretion, delays gastric emptying, and acts as a satiety agent.
Tekniske data
Renhed 96.9%
CAS 122384-88-7
Indhold og opbevaring Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Til brug med (applikation) Metabolism-sugar/lipid metabolism
Molekylvægt (g/mol) 3903.28 g/mol
Produkttype Peptides
Mængde 1 mg
Forsendelsestilstand Room temperature
Synonym DAP amide, human