Få mere at vide
MedChemExpress Amylin (8-37), rat
Artikelnummer. 16472108
Encompass_Preferred
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
16472108 5 mg
1 options
Artikelnummer. 16472108 Leverandør MedChemExpress Leverandørnr. HYP1473
MedChemExpress Amylin (8-37), rat is a truncated analog of native Amylin that selectively inhibits insulin-related glucose uptake and glycogen deposition in muscle tissue. Amylin (8-37), rat is a weak amylin receptor (AMY) antagonist.
Tekniske data
Renhed 99.88%
CAS 138398-61-5
Indhold og opbevaring Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Til brug med (applikation) Metabolism-sugar/lipid metabolism
Molekylvægt (g/mol) 3200.61 g/mol
Produkttype Peptides
Mængde 5 mg
Forsendelsestilstand Room temperature
Opløselighed H2O : 50 mg/mL (15.62 mM∣Need ultrasonic)
Synonym Amylin (8-37) (mouse, rat)