Få mere at vide
Amygdalin, MedChemExpress
Artikelnummer. 16491496
Encompass_Preferred
Skift visning
Mængde:
50 mg
100 mg
200 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16491496 200 mg
16459026 100 mg
16424787 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16491496 Leverandør Medchem Express Leverandørnr. HYN0190
MedChemExpress Amygdalin is a plant glucoside isolated from the stones of rosaceous fruits, such as apricots, peaches, almond, cherries, and plums.
Amygdalin is a plant glucoside isolated from the stones of rosaceous fruits, such as apricots, peaches, almond, cherries, and plums.
Tekniske data
Kemisk navn eller materiale Amygdalin
CAS 29883-15-6
Farve White
Renhedsgrad noter Research
Molekylær formel C20H27NO11
Mængde 200 mg
Opløselighedsinformation DMSO : ≥ 75 mg/mL (163.96 mM) ∣H2O : 50 mg/mL (109.31 mM; Need ultrasonic)
SMIL O[C@H]1[C@H](O[C@@H](C#N)C2=CC=CC=C2)O[C@H](CO[C@H]3[C@@H]([C@H]([C@@H]([C@@H](CO)O3)O)O)O)[C@@H](O)[C@@H]1O
Molekylvægt (g/mol) 457.43
Formel vægt 457.43
Procent renhed 99.52%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death