Få mere at vide
Amprotropine Phosphate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Amprotropine Phosphate |
| CAS | 134-53-2 |
| Analyt- eller komponentnavne | Amprotropine Phosphate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H29 N O3 . H3 O4 P |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 3-(Diethylamino)-2,2-dimethylpropyl 3-hydroxy-2-phenylpropanoate phosphate (1:1), Amprotropine phosphate, Syntropan, 3-Diethylamino-2,2-dimethylpropyl Tropate Phosphate, Amprotropine phosphate (6CI), Tropic acid, 3-(diethylamino)-2,2-dimethylpropyl ester, phosphate (1:1) (8CI), Tropic acid, 3-(diethylamino)-2,2-dimethylpropyl ester, phosphate (7CI), 1-Propanol, 3-(diethylamino)-2,2-dimethyl-, tropate, phosphate (8CI), AP 407, Syntropan, Benzeneacetic acid, α-(hydroxymethyl)-, 3-(diethylamino)-2,2-dimethylpropyl ester, phosphate (1:1) (salt) (9CI) |
| InChI formel | InChI=1S/C18H29NO3.H3O4P/c1-5-19(6-2)13-18(3,4)14-22-17(21)16(12-20)15-10-8-7-9-11-15;1-5(2,3)4/h7-11,16,20H,5-6,12-14H2,1-4H3;(H3,1,2,3,4) |
| Løsningstype | Neat |
| SMIL | CCN(CC)CC(C)(C)COC(=O)C(CO)c1ccccc1.OP(=O)(O)O |
| Vis mere |