Få mere at vide
Amprenavir-d4, TRC
Artikelnummer. 30341551
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30341551 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30341551 Leverandør Toronto Research Chemicals Leverandørnr. TRCA63440210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Amprenavir-d4
CAS 1217661-20-5
Molekylær formel C25H31D4N3O6S
Mængde 10 mg
Synonym 141W94-d4, KVX-478-d4, Agenerase-d4, Prozei-d4
InChI formel InChI=1S/C25H35N3O6S/c1-18(2)15-28(35(31,32)22-10-8-20(26)9-11-22)16-24(29)23(14-19-6-4-3-5-7-19)27-25(30)34-21-12-13-33-17-21/h3-11,18,21,23-24,29H,12-17,26H2,1-2H3,(H,27,30)/t21-,23-,24+/m0/s1/i13D2,17D2
SMIL O=C(N[C@H]([C@@H](CN(S(C1=CC=C(N)C=C1)(=O)=O)CC(C)C)O)CC2=CC=CC=C2)O[C@H]3CC([2H])([2H])OC3([2H])[2H]
IUPAC navn [(3S)-2,2,5,5-tetradeuteriooxolan-3-yl] N-[(2S,3R)-4-[(4-aminophenyl)sulfonyl-(2-methylpropyl)amino]-3-hydroxy-1-phenylbutan-2-yl]carbamate
Molekylvægt (g/mol) 509.65