Learn More
Simport™ Scientific Amplate™ T323 Series Thin Wall PCR Plate
Available with black alpha-numeric lettering. Simport™ Scientific Amplate™ T323 Series Thin Wall PCR Plate, each well features ultrathin sidewalls for uniform and efficient temperature transfer.
POA
Specifications
Sterility | Non-sterile |
---|---|
No. of Wells | 48 |
Material | Polypropylene |
No. per Case | 50 |
Description
- Made of polypropylene
- Available with black alpha-numeric
- Flexible, can be cut to desired format
- Autoclavable
- Inside of tube is smooth and has an inert surface
Certifications
RNase, DNase, Pyrogen and DNA-free
Specifications
Non-sterile | |
48 | |
Polypropylene | |
50 |