Learn More
Simport™ Scientific Amplate™ T323 Series Thin Wall PCR Plate
Available with black alpha-numeric lettering. Simport™ Scientific Amplate™ T323 Series Thin Wall PCR Plate, each well features ultrathin sidewalls for uniform and efficient temperature transfer.
POA
Specifications
Sterility Non-sterile
No. of Wells 48
Material Polypropylene
No. per Case 50
View More Specs
Description
Description
  • Made of polypropylene
  • Available with black alpha-numeric
  • Flexible, can be cut to desired format
  • Autoclavable
  • Inside of tube is smooth and has an inert surface
Certifications
RNase, DNase, Pyrogen and DNA-free
Specifications
Specifications
Non-sterile
48
Polypropylene
50