Learn More
Simport™ Scientific Amplate™ T323 Series Thin Wall PCR Plate
Available with black alpha-numeric lettering. Simport™ Scientific Amplate™ T323 Series Thin Wall PCR Plate, each well features ultrathin sidewalls for uniform and efficient temperature transfer.
Brand:  Simport™ Scientific T323-24Y
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11516699
  • 342.00 DKK / Pack of 50
Estimated Shipment
29-06-2021
Add to basket
Description
Description
  • Made of polypropylene
  • Available with black alpha-numeric
  • Flexible, can be cut to desired format
  • Autoclavable
  • Inside of tube is smooth and has an inert surface
Certifications
RNase, DNase, Pyrogen and DNA-free
Specifications
Specifications
Autoclavable
Polypropylene
50/Cs.
24
Standard
Yellow
50
Non-sterile
4 x 6