Learn More
Invitrogen™ AMPK (A1/B2/G1) Recombinant Human Protein
AMPK (A1/B2/G1) is a heterotrimeric protein consisting of an α catalytic subunit, and non-catalytic β and γ subunits. Recombinant full-length human AMPK (combination of A1/B2/G1 subunits) was expressed by baculovirus in Sf9 insect cells using a C-terminal His tags.
Brand: Invitrogen™ PV6244
Additional Details : Weight : 0.01000kg
Description
AMPKs are an important energy-sensing enzyme group in the cells that monitor energy status particularly in response to stress. AMPKs regulate fatty acid and cholesterol synthesis by regulating the key rate-limiting enzymes acetyl-CoA carboxylase and hydroxy beta-methylglutaryl-CoA reductase. The beta subunit may be a positive regulator of AMPK activity and is highly expressed in skeletal muscle.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation | |
63.6/31.4/ 38.7kDa | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. | |
aa 10-end | |
Recombinant |
50mM sodium phosphate pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.2mM DTT, 25% glycerol | |
10μg | |
Subunit A1: PRKAA1, MGC33776, MGC57364 Subunit B2: PRKAB2, MGC61468 Subunit G1: PRKAG1, AMPKG, MGC8666 | |
Human | |
His Tag |